Assay Details
| Gene Symbol: MB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214236.1 | NP_999401.1 | 451 | 95 | ||
| GenBank mRNA | AK396342.1 | - | 461 | 95 | ||
| M14433.1 | - | 451 | 95 | |||
More Information
Set Membership: |
|

MB
Genomic Map