Assay Details
| Gene Symbol: FSHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214386.3 | NP_999551.2 | 10 | 2048 | 99 | |
| XM_021085881.1 | XP_020941540.1 | 2113 | 99 | |||
| XM_021085882.1 | XP_020941541.1 | 2041 | 99 | |||
| XM_021085883.1 | XP_020941542.1 | 1967 | 99 | |||
| XM_021085884.1 | XP_020941543.1 | 1931 | 99 | |||
| GenBank mRNA | AF025377.1 | - | 10 | 2048 | 99 | |
More Information
Set Membership: |
|

FSHR
Genomic Map