Assay Details
| Gene Symbol: C8G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097452.1 | NP_001090921.1 | 551 | 62 | ||
| XM_005652714.3 | XP_005652771.1 | 914 | 62 | |||
| XM_013989044.2 | XP_013844498.1 | 966 | 62 | |||
| XM_013989048.2 | XP_013844502.1 | 937 | 62 | |||
| GenBank mRNA | AK232757.1 | - | 552 | 62 | ||
| AK233484.1 | - | 1052 | 62 | |||
| AK233683.1 | - | 1042 | 62 | |||
| DQ333202.1 | - | 551 | 62 | |||
More Information
Set Membership: |
|

C8G
Genomic Map