Assay Details
| Gene Symbol: DESI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097419.1 | NP_001090888.1 | 2 - 3 | 357 | 85 | |
| XM_005667956.3 | XP_005668013.1 | 364 | 85 | |||
| XM_005667957.3 | XP_005668014.1 | 598 | 85 | |||
| XM_005667960.3 | XP_005668017.1 | 215 | 85 | |||
| XM_013980072.2 | XP_013835526.1 | 379 | 85 | |||
| XM_013980075.2 | XP_013835529.1 | 377 | 85 | |||
| GenBank mRNA | DQ435075.1 | - | 2 - 3 | 357 | 85 | |
More Information
Set Membership: |
|

DESI2
Genomic Map