Assay Details
| Gene Symbol: ROMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097462.2 | NP_001090931.2 | 2 - 3 | 194 | 139 | |
| GenBank mRNA | AK347158.1 | - | 2 - 3 | 175 | 139 | |
| AK392584.1 | - | 2 - 3 | 209 | 139 | ||
| AK400934.1 | - | 504 | 139 | |||
| DQ629137.1 | - | 2 - 3 | 194 | 139 | ||
| EV894502.2 | - | 2 - 3 | 216 | 139 | ||
More Information
Set Membership: |
|

ROMO1
Genomic Map