Assay Details
| Gene Symbol: BMP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105290.1 | NP_001098760.1 | 2 - 3 | 612 | 71 | |
| XM_005673044.3 | XP_005673101.1 | 1165 | 71 | |||
| XM_005673045.3 | XP_005673102.1 | 1167 | 71 | |||
| XM_021077319.1 | XP_020932978.1 | 1167 | 71 | |||
| XM_021077320.1 | XP_020932979.1 | 1167 | 71 | |||
| GenBank mRNA | AK351088.1 | - | 2 - 3 | 1010 | 71 | |
| AY773017.1 | - | 2 - 3 | 582 | 71 | ||
More Information
Set Membership: |
|

BMP7
Genomic Map