Assay Details
Gene Symbol: MUC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113440.1 | NP_001106911.1 | 3 - 4 | 1551 | 65 | |
GenBank mRNA | EU330422.1 | - | 3 - 4 | 1551 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
