Assay Details
| Gene Symbol: MDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123138.1 | NP_001116610.1 | 13 - 14 | 5959 | 61 | |
| XM_005665729.3 | XP_005665786.1 | 6098 | 61 | |||
| XM_013977586.2 | XP_013833040.1 | 6074 | 61 | |||
| XM_013977587.2 | XP_013833041.1 | 6001 | 61 | |||
| XM_013977588.2 | XP_013833042.1 | 6260 | 61 | |||
| XM_013977589.2 | XP_013833043.1 | 6113 | 61 | |||
| XM_013977590.2 | XP_013833044.1 | 6093 | 61 | |||
| XM_013977591.2 | XP_013833045.1 | 5984 | 61 | |||
| XM_013977592.2 | XP_013833046.1 | 6117 | 61 | |||
| XM_021097919.1 | XP_020953578.1 | 6077 | 61 | |||
| XM_021097920.1 | XP_020953579.1 | 6252 | 61 | |||
| XM_021097922.1 | XP_020953581.1 | 5969 | 61 | |||
| XM_021097923.1 | XP_020953582.1 | 6160 | 61 | |||
| GenBank mRNA | AK392334.1 | - | 14 - 15 | 5955 | 61 | |
More Information
Set Membership: |
|

MDC1
Genomic Map