Assay Details
Gene Symbol: WDR83OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001645.1 | NP_001001645.1 | 2 - 3 | 187 | 104 | |
XM_005652849.2 | XP_005652906.1 | 313 | 104 | |||
GenBank mRNA | AK399947.1 | - | 2 - 3 | 204 | 104 | |
AY553921.1 | - | 2 - 3 | 187 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
