Assay Details
| Gene Symbol: FCGR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033011.1 | NP_001028183.1 | 5 - 6 | 843 | 66 | |
| GenBank mRNA | AK345322.1 | - | 5 - 6 | 843 | 66 | |
| AK345770.1 | - | 5 - 6 | 869 | 66 | ||
| DQ026063.1 | - | 5 - 6 | 843 | 66 | ||
| KF595281.1 | - | 5 - 6 | 842 | 66 | ||
| KF595282.1 | - | 5 - 6 | 791 | 66 | ||
More Information
Set Membership: |
|

FCGR1A
Genomic Map