Assay Details
| Gene Symbol: SLPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213870.1 | NP_999035.1 | 3 - 4 | 385 | 73 | |
| GenBank mRNA | M57446.1 | - | 3 - 4 | 385 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SLPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213870.1 | NP_999035.1 | 3 - 4 | 385 | 73 | |
| GenBank mRNA | M57446.1 | - | 3 - 4 | 385 | 73 | |
Set Membership: |
|