Assay Details
| Gene Symbol: EDN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213882.1 | NP_999047.1 | 2 - 3 | 308 | 61 | |
| GenBank mRNA | AK348522.1 | - | 2 - 3 | 568 | 61 | |
| X07383.1 | - | 308 | 61 | |||
More Information
Set Membership: |
|

EDN1
Genomic Map