Assay Details
| Gene Symbol: VIPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214036.1 | NP_999201.1 | 11 | 1256 | 71 | |
| XM_005669395.3 | XP_005669452.1 | 1236 | 71 | |||
| XM_005669397.3 | XP_005669454.1 | 1008 | 71 | |||
| XM_013981507.2 | XP_013836961.1 | 1155 | 71 | |||
| XM_013981508.2 | XP_013836962.1 | 1235 | 71 | |||
| XM_013981510.2 | XP_013836964.1 | 756 | 71 | |||
| XM_021068242.1 | XP_020923901.1 | 1896 | 71 | |||
| XM_021068243.1 | XP_020923902.1 | 1123 | 71 | |||
| GenBank mRNA | HM209403.1 | - | 10 | 1014 | 71 | |
| HM209404.1 | - | 8 | 776 | 71 | ||
| HM209405.1 | - | 10 | 904 | 71 | ||
| HM209406.1 | - | 12 | 1235 | 71 | ||
| HM209407.1 | - | 10 | 1013 | 71 | ||
| HM209408.1 | - | 8 | 756 | 71 | ||
| HM209409.1 | - | 8 | 744 | 71 | ||
| HM209410.1 | - | 7 | 667 | 71 | ||
| HM209411.1 | - | 7 | 647 | 71 | ||
| U49434.2 | - | 11 | 1256 | 71 | ||
More Information
Set Membership: |
|

VIPR1
Genomic Map