Assay Details
| Gene Symbol: HCST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214142.1 | NP_999307.1 | 2 - 3 | 168 | 115 | |
| XM_013998329.2 | XP_013853783.1 | 322 | 115 | |||
| XM_013998330.2 | XP_013853784.1 | 322 | 115 | |||
| XM_013998331.2 | XP_013853785.1 | 320 | 115 | |||
| XM_013998332.2 | XP_013853786.1 | 295 | 115 | |||
| GenBank mRNA | AF285444.1 | - | 2 - 3 | 168 | 115 | |
| AF285445.1 | - | 2 - 3 | 74 | 115 | ||
| AK399783.1 | - | 2 - 3 | 126 | 115 | ||
More Information
Set Membership: |
|

HCST
Genomic Map