Assay Details
| Gene Symbol: HCST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214142.1 | NP_999307.1 | 3 - 4 | 298 | 109 | |
| XM_013998329.2 | XP_013853783.1 | 452 | 109 | |||
| XM_013998331.2 | XP_013853785.1 | 450 | 109 | |||
| XM_013998332.2 | XP_013853786.1 | 425 | 109 | |||
| GenBank mRNA | AF285444.1 | - | 3 - 4 | 298 | 109 | |
| AF285445.1 | - | 3 - 4 | 204 | 109 | ||
| AK399783.1 | - | 3 - 4 | 256 | 109 | ||
More Information
Set Membership: |
|

HCST
Genomic Map