Assay Details
| Gene Symbol: CYP51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214432.1 | NP_999597.1 | 5 - 6 | 781 | 75 | |
| XM_005667583.3 | XP_005667640.1 | 757 | 75 | |||
| XM_005667584.3 | XP_005667641.1 | 676 | 75 | |||
| GenBank mRNA | AB009988.1 | - | 5 - 6 | 781 | 75 | |
| AB529453.1 | - | 5 - 6 | 754 | 75 | ||
| AF198112.1 | - | 3 - 4 | 475 | 75 | ||
| AK235908.1 | - | 5 - 6 | 912 | 75 | ||
| AK394700.1 | - | 3 - 4 | 327 | 75 | ||
| AK397262.1 | - | 5 - 6 | 935 | 75 | ||
| AY609713.1 | - | 5 - 6 | 879 | 75 | ||
More Information
Set Membership: |
|

CYP51
Genomic Map