Assay Details
| Gene Symbol: CALB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130226.1 | NP_001123698.1 | 5 - 6 | 375 | 107 | |
| GenBank mRNA | AK393398.1 | - | 5 - 6 | 605 | 107 | |
| EU743962.1 | - | 5 - 6 | 375 | 107 | ||
More Information
Set Membership: |
|

CALB1
Genomic Map