Assay Details
| Gene Symbol: ZBTB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101821.1 | NP_001095291.1 | 2 | 1352 | 71 | |
| XM_013977632.2 | XP_013833086.1 | 1220 | 71 | |||
| GenBank mRNA | EU282372.1 | - | 2 | 1166 | 71 | |
More Information
Set Membership: |
|

ZBTB12
Genomic Map