Assay Details
| Gene Symbol: HAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136212.1 | NP_001129684.1 | 4 | 1947 | 93 | |
| GenBank mRNA | AB377191.1 | - | 4 | 1947 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: HAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136212.1 | NP_001129684.1 | 4 | 1947 | 93 | |
| GenBank mRNA | AB377191.1 | - | 4 | 1947 | 93 | |
Set Membership: |
|