Assay Details
| Gene Symbol: SGCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144123.1 | NP_001137595.1 | 8 | 844 | 79 | |
| XM_021076526.1 | XP_020932185.1 | 1236 | 79 | |||
| XM_021076527.1 | XP_020932186.1 | 1641 | 79 | |||
| GenBank mRNA | AY609606.1 | - | 8 | 952 | 79 | |
| FJ481169.1 | - | 8 | 844 | 79 | ||
More Information
Set Membership: |
|

SGCD
Genomic Map