Assay Details
| Gene Symbol: AGPAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130534.1 | NP_001124006.1 | 787 | 56 | ||
| GenBank mRNA | AB527060.1 | - | 661 | 56 | ||
| AK232197.1 | - | 766 | 56 | |||
| AK346049.1 | - | 763 | 56 | |||
| AK391026.1 | - | 771 | 56 | |||
| AK399232.1 | - | 800 | 56 | |||
| EU780066.1 | - | 787 | 56 | |||
| FJ439665.1 | - | 662 | 56 | |||
More Information
Set Membership: |
|

AGPAT2
Genomic Map