Assay Details
| Gene Symbol: TCF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167591.1 | NP_001161063.1 | 1 - 2 | 236 | 82 | |
| XM_005665754.3 | XP_005665811.1 | 774 | 82 | |||
More Information
Set Membership: |
|

TCF19
Genomic Map