Assay Details
| Gene Symbol: TCF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167591.1 | NP_001161063.1 | 2 - 3 | 804 | 71 | |
| XM_005665754.3 | XP_005665811.1 | 1342 | 71 | |||
| XM_013977603.2 | XP_013833057.1 | 1402 | 71 | |||
More Information
Set Membership: |
|

TCF19
Genomic Map