Assay Details
Gene Symbol: ELOVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167634.1 | NP_001161106.1 | 3 - 4 | 387 | 67 | |
XM_005671398.2 | XP_005671455.1 | 482 | 67 | |||
GenBank mRNA | AB529460.1 | - | 3 - 4 | 387 | 67 | |
AK238786.1 | - | 3 - 4 | 482 | 67 | ||
AK351721.1 | - | 3 - 4 | 482 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
