Assay Details
| Gene Symbol: IVD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167635.1 | NP_001161107.1 | 4 - 5 | 466 | 62 | |
| GenBank mRNA | AB529855.1 | - | 4 - 5 | 466 | 62 | |
| AK238259.1 | - | 4 - 5 | 494 | 62 | ||
| AK240462.1 | - | 4 - 5 | 469 | 62 | ||
| AK345817.1 | - | 4 - 5 | 469 | 62 | ||
| AK393269.1 | - | 4 - 5 | 502 | 62 | ||
More Information
Set Membership: |
|

IVD
Genomic Map