Assay Details
| Gene Symbol: HSD17B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167649.1 | NP_001161121.1 | 3 - 4 | 667 | 57 | |
| GenBank mRNA | AB529535.1 | - | 3 - 4 | 667 | 57 | |
| AK233331.1 | - | 3 - 4 | 868 | 57 | ||
More Information
Set Membership: |
|

HSD17B2
Genomic Map