Assay Details
Gene Symbol: TRIM39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128479.1 | NP_001121951.1 | 1 - 2 | 451 | 64 | |
XM_005653455.3 | XP_005653512.1 | 553 | 64 | |||
XM_013996585.2 | XP_013852039.1 | 888 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
