Assay Details
| Gene Symbol: CLDN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160081.1 | NP_001153553.1 | 3 - 4 | 646 | 63 | |
| XM_021068338.1 | XP_020923997.1 | 559 | 63 | |||
| GenBank mRNA | FJ875103.1 | - | 3 - 4 | 646 | 63 | |
More Information
Set Membership: |
|

CLDN18
Genomic Map