Assay Details
| Gene Symbol: SPATA19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177920.1 | NP_001171391.1 | 6 - 7 | 556 | 71 | |
| GenBank mRNA | GU475012.1 | - | 6 - 7 | 556 | 71 | |
More Information
Set Membership: |
|

SPATA19
Genomic Map