Assay Details
Gene Symbol: TOMM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190159.1 | NP_001177088.1 | 5 - 6 | 810 | 77 | |
XM_005672959.3 | XP_005673016.1 | 855 | 77 | |||
XM_005672960.3 | XP_005673017.1 | 726 | 77 | |||
GenBank mRNA | AK239658.1 | - | 5 - 6 | 810 | 77 | |
AK347559.1 | - | 5 - 6 | 752 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
