Assay Details
| Gene Symbol: MBOAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190423.1 | NP_001177352.1 | 2 - 3 | 414 | 85 | |
| GenBank mRNA | HM070295.1 | - | 2 - 3 | 414 | 85 | |
More Information
Set Membership: |
|

MBOAT4
Genomic Map