Assay Details
| Gene Symbol: MOSPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195354.1 | NP_001182283.1 | 2 - 3 | 243 | 70 | |
| XM_005654294.3 | XP_005654351.1 | 70 | ||||
| XM_013983792.2 | XP_013839246.1 | 70 | ||||
| GenBank mRNA | GU373668.1 | - | 2 - 3 | 243 | 70 | |
More Information
Set Membership: |
|

MOSPD1
Genomic Map