Assay Details
| Gene Symbol: PALM2AKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204181.2 | NP_001191110.1 | 1 - 2 | 137 | 85 | |
| GenBank mRNA | AK348182.1 | - | 1 - 2 | 136 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001204181.2 transcripts.
Set Membership: |
|

PALM2AKAP2
Genomic Map