Assay Details
| Gene Symbol: FLVCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142840.1 | NP_001136312.1 | 1 - 2 | 877 | 61 | |
| GenBank mRNA | AK389329.1 | - | 1 - 2 | 336 | 61 | |
| FJ236708.1 | - | 1 - 2 | 877 | 61 | ||
| FJ436993.1 | - | 1 - 2 | 740 | 61 | ||
More Information
Set Membership: |
|

FLVCR2
Genomic Map