Assay Details
| Gene Symbol: FASTK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143707.2 | NP_001137179.1 | 7 - 8 | 1335 | 53 | |
| XM_005657686.3 | XP_005657743.1 | 1645 | 53 | |||
| XM_013990725.1 | XP_013846179.1 | 1788 | 53 | |||
| XM_013990726.1 | XP_013846180.1 | 1767 | 53 | |||
| XM_021078454.1 | XP_020934113.1 | 1387 | 53 | |||
| XM_021078455.1 | XP_020934114.1 | 1618 | 53 | |||
| GenBank mRNA | AK350225.1 | - | 7 - 8 | 1339 | 53 | |
| AK397083.1 | - | 7 - 8 | 1383 | 53 | ||
| FJ438466.1 | - | 7 - 8 | 1338 | 53 | ||
More Information
Set Membership: |
|

FASTK
Genomic Map