Assay Details
Gene Symbol: TBCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205403.1 | NP_001192332.1 | 4 - 5 | 628 | 105 | |
GenBank mRNA | AK240439.1 | - | 4 - 5 | 620 | 105 | |
AK352168.1 | - | 4 - 5 | 601 | 105 | ||
AK397066.1 | - | 994 | 105 | |||
CJ016107.1 | - | 4 - 5 | 628 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
