Assay Details
Gene Symbol: RAB11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244875.1 | NP_001231804.1 | 1 - 2 | 435 | 61 | |
GenBank mRNA | AK348825.1 | - | 1 - 2 | 437 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
