Assay Details
| Gene Symbol: LAYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244894.1 | NP_001231823.1 | 1 - 2 | 254 | 92 | |
| GenBank mRNA | FD642694.1 | - | 1 - 2 | 259 | 92 | |
More Information
Set Membership: |
|

LAYN
Genomic Map