Assay Details
| Gene Symbol: LAYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244894.1 | NP_001231823.1 | 5 - 6 | 823 | 69 | |
| GenBank mRNA | AK240128.1 | - | 4 - 5 | 522 | 69 | |
More Information
Set Membership: |
|

LAYN
Genomic Map