Assay Details
| Gene Symbol: YIPF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001245980.1 | NP_001232909.1 | 6 - 7 | 803 | 71 | |
| XM_005666032.3 | XP_005666089.1 | 917 | 71 | |||
| XM_005666033.3 | XP_005666090.1 | 923 | 71 | |||
| GenBank mRNA | AK234085.1 | - | 6 - 7 | 717 | 71 | |
| AK235735.1 | - | 6 - 7 | 854 | 71 | ||
| AK236195.1 | - | 6 - 7 | 822 | 71 | ||
| AK389571.1 | - | 6 - 7 | 896 | 71 | ||
| AK394357.1 | - | 6 - 7 | 822 | 71 | ||
| AY609788.1 | - | 6 - 7 | 801 | 71 | ||
| EU559702.1 | - | 6 - 7 | 803 | 71 | ||
More Information
Set Membership: |
|

YIPF3
Genomic Map