Assay Details
| Gene Symbol: CPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246243.1 | NP_001233172.1 | 1 - 2 | 616 | 81 | |
| GenBank mRNA | GU929943.1 | - | 1 - 2 | 616 | 81 | |
More Information
Set Membership: |
|

CPT2
Genomic Map