Assay Details
| Gene Symbol: ETV5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243025.1 | NP_001229954.1 | 1 - 2 | 153 | 84 | |
| XM_021068467.1 | XP_020924126.1 | 288 | 84 | |||
| GenBank mRNA | AK232501.1 | - | 1 - 2 | 153 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001243025.1 transcripts.
Set Membership: |
|

ETV5
Genomic Map