Assay Details
| Gene Symbol: TMEM61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243365.1 | NP_001230294.1 | 1 - 2 | 214 | 82 | |
| GenBank mRNA | AK231183.1 | - | 1 - 2 | 214 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001243365.1 transcripts.
Set Membership: |
|

TMEM61
Genomic Map