Assay Details
| Gene Symbol: TMEM61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243365.1 | NP_001230294.1 | 3 - 4 | 623 | 75 | |
| GenBank mRNA | AK231183.1 | - | 3 - 4 | 623 | 75 | |
More Information
Set Membership: |
|

TMEM61
Genomic Map