Assay Details
| Gene Symbol: SNAPIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243486.1 | NP_001230415.1 | 3 - 4 | 316 | 74 | |
| XM_005663352.3 | XP_005663409.1 | 360 | 74 | |||
| GenBank mRNA | AK348954.1 | - | 3 - 4 | 313 | 74 | |
| AY610287.1 | - | 3 - 4 | 321 | 74 | ||
| EW258917.2 | - | 1 - 2 | 113 | 74 | ||
More Information
Set Membership: |
|

SNAPIN
Genomic Map