Assay Details
Gene Symbol: GSKIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243496.1 | NP_001230425.1 | 2 - 3 | 272 | 85 | |
XM_005666425.3 | XP_005666482.1 | 439 | 85 | |||
GenBank mRNA | AK230979.1 | - | 2 - 3 | 274 | 85 | |
AK389717.1 | - | 2 - 3 | 297 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
