Assay Details
| Gene Symbol: ABHD17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243558.1 | NP_001230487.1 | 2 - 3 | 1040 | 101 | |
| XM_005660158.3 | XP_005660215.1 | 1185 | 101 | |||
| GenBank mRNA | AK349239.1 | - | 2 - 3 | 985 | 101 | |
More Information
Set Membership: |
|

ABHD17B
Genomic Map