Assay Details
| Gene Symbol: NHLRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243561.1 | NP_001230490.1 | 4 - 5 | 937 | 67 | |
| GenBank mRNA | AK347629.1 | - | 4 - 5 | 939 | 67 | |
More Information
Set Membership: |
|

NHLRC2
Genomic Map