Assay Details
| Gene Symbol: P2RY10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243677.1 | NP_001230606.1 | 1 - 2 | 57 | 95 | |
| XM_005673737.3 | XP_005673794.1 | 120 | 95 | |||
| GenBank mRNA | AK238879.1 | - | 1 - 2 | 57 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001243677.1, XM_005673737.3 transcripts.
Set Membership: |
|

P2RY10
Genomic Map