Assay Details
| Gene Symbol: CCDC59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243859.1 | NP_001230788.1 | 3 - 4 | 602 | 98 | |
| GenBank mRNA | AK240595.1 | - | 3 - 4 | 602 | 98 | |
More Information
Set Membership: |
|

CCDC59
Genomic Map